Synthware Vacuum Distilling Adapter, Kemtech America
Catalog # 60000-700
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- ST Joint:14/20
- Description:Synthware Vacuum Distilling Adapter, 8 mm hose O.D.
- Cat. no.:60000-700
- Supplier No.:A491420
Specifications
About this item
This vacuum distillation adapter is useful in the sub-boiling separation of mixtures with volatile components.
- 100mm span between joints
- [ST] joints
The span between joints is 100 mm.