Plunger Cans, Justrite®
Supplier: Justrite
Certificates
About this item
Plunger can helps moisten cleaning cloths or swabs quickly and easily
Upper pan fills when pushed down, and surplus liquid drains safely back into reservoir. Perforated pan screen serves as a flame arrester. Top rims have molded edge protectors for smooth, trouble-free moistening. Brass pump assembly. Available in terne plate or high-density polyethylene construction.
Certifications: FM approved.
Customers Who Bought This Also Bought