InVitro™ Biotainer® Carboys, PETG, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Designed for pharmaceutical and biological applications.
- Individually wrapped within case
- Sterilized by gamma irradiation (SAL 10⁻⁶)
- Caps are white polypropylene with low-density polyethylene foam liner
Carboys are manufactured from transparent PETG, which features high impact strength, good chemical resistance, and excellent gas barrier properties.
Neck size: 48 mm (Double Start Thread)
Certifications: Carboys meet USP Class VI Standards, and are nonpyrogenic and noncytotoxic.
Customers Who Bought This Also Bought