PolyGen™ Mops, Micronova
Supplier: Micronova
Certificates
About this item
PolyGen™ mops are ideal for disinfecting in pharmaceutical and biotech environments.
- Low-linting polyester looped strands with dual-action abrasive mesh
- Herringbone weave ensures there are no raw edges, and limits particle generation
- Mops are reuseable, launderable, and autoclavable
LoopMop™ mops are knitted polyester looped mops for applying disinfectant in pharmaceutical, aseptic processing, and medical device applications. The polyester material is absorbent enough to clean to a dry finish but will leave enough moisture for disinfectants that require an extended drying time. LoopMop™ mops are reuseable, launderable, and autoclavable.
All mop heads attach to the stainless steel universal handle by means of the orange polypropylene quick-disconnect adapter. For less critical applications, choose the fiberglass/polypropylene handle with attached adapter (68400-002).
Ordering information: All components are ordered separately. For additional handle lengths, see the 68400-302 series.
Specifications
- Catalog No:
- 68400-786
- 68399-706
- 68400-758
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
Specifications
Related Information
Certificates
Search by Lot NumberED-Micronova- Green Cleaning
Additional Product Details
Focus Safety Volume 1 Issue 1
Product Literature
Frequently Bought Together