Kimtech™ G3 Sapphire Nitrile Cleanroom Gloves
Supplier: Microflex
Certificates
About this item
Sapphire blue nitrile cleanroom gloves protect against contamination for products and processes in cleanroom manufacturing.
- Powder-free
- Straight fingers
- Ambidestrious
- Textured fingertip
- Beaded cuff
- Double chlorinated
- Laundered
- Bulk packaged
Gloves are designed for cleanroom/industrial use only. They meet FDA food contact requirements set forth in 21 CFR 177.2600.
Packaging: Gloves are packaged 100 per pack, 1000 gloves per case.
Specifications
- Catalog No:
- 76575-514
- 76575-508
- 76575-512
- 76575-516
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Finger thickness:
- 7.1 mil
- 7.1 mil
- 7.1 mil
- 7.1 mil
- Palm thickness:
- 5.5 mil
- 5.5 mil
- 5.5 mil
- 5.5 mil