Order Entry
Canada
ContactUsLinkComponent
VeriSpec® Multi-Element Standard 24, Ricca Chemicals
VeriSpec® Multi-Element Standard 24, Ricca Chemicals
Catalog # RCRV010714100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec® Multi-Element Standard 24, Ricca Chemicals
Catalog # RCRV010714100N
Supplier:  Ricca Chemical
Supplier Number:  RV010714-100N
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃/tr. HCl
  • Pack type:
    natural LDPE
  • Description:
    VeriSpec® Multi-Element Standard 24
  • Size:
    100 ml
  • Cat. no.:
    RCRV010714100N
  • Shelf Life:
    36 months
  • Supplier No.:
    RV010714-100N

Specifications

About this item

100 ppm Al, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, Ga, K, Li, Mg, Mn, Na, Ni, Pb, Se, Sr, Te, Tl, Zn

Certifications: Manufactured and Tested in an ISO 17025 accredited laboratory.