ContactUsLinkComponent
ZymoBIOMICS Lysis Solution, Zymo Research
ZymoBIOMICS Lysis Solution, Zymo Research
  76358-806
 :  
undefined
ZymoBIOMICS Lysis Solution, Zymo Research
  76358-806
 :  D4300-1-150
 :  
  • Description:
    ZymoBIOMICS Lysis Solution
  • Size:
    150 ml
  • Cat. no.:
    76358-806
ZymoBIOMICS Lysis Solution is designed to be used with the ZymoBIOMICS DNA extraction kits. Used in combination with the ZR BashingBead Lysis Tubes, it assists with microbial lysis. It is certified to be low-bioburden to ensure minimal background contaminants for low abundance microbiome samples.