Order Entry
Canada
ContactUsLinkComponent
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (RPE-Astral™616)
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (RPE-Astral™616)
Supplier:  Biotium
Certificates
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (RPE-Astral™616)
Supplier:  Biotium

Specifications

  • Catalog No:
  • 77449-082
  • 77449-084
  • Antigen symbol:
  • CD19
  • CD19
  • Antigen synonyms:
  • T-cell Surface Antigen Leu-12|CVID3|B-Lymphocytes Surface Antigen B4|B4
  • T-cell Surface Antigen Leu-12|CVID3|B-Lymphocytes Surface Antigen B4|B4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • RPE (R-Phycoerythrin)
  • RPE (R-Phycoerythrin)
  • Clone:
  • SJ25C1
  • SJ25C1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 930
  • 930
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Molecular weight:
  • 95 kDa
  • 95 kDa
  • Concentration:
  • 50 µg/ml
  • 50 µg/ml
  • Storage temperature:
  • Store at 2... 8 °C, protect fluorescent conjugates from light
  • Store at 2... 8 °C, protect fluorescent conjugates from light
  • Shipping temperature:
  • RT
  • RT

Specifications

Frequently Bought Together