Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
Home
Inquarts, Herman
Herman Inquarts,, pellets
Herman Inquarts,, pellets
Herman Inquarts,, pellets
Catalog # 23923-006
Supplier:
Bico
CAS Number:
Expand image
Herman Inquarts,, pellets
Catalog # 23923-006
Supplier:
Bico
Supplier Number:
239-23
CAS Number:
Specifications
Size:
1000 pellets
Cat. no.:
23923-006
Supplier No.:
239-23
Specifications
About this item
Product Details
:
Product Details