SP Wilmad-LabGlass Precision Bore Class A Burettes with PTFE Stopcock, SP Industries
Supplier: SP Industries
Certificates
About this item
Made of borosilicate glass, these precision type, Class A burets have a straight 2mm PTFE stopcock
Calibration is in compliance with ASTM E 694 specifications.
Certifications: Reference No. ASTM E 287 Laboratory Glass Graduated Burets.
Related Information
Bel-Art_New_Product_Supplement_Laboratory_Problem_Solvers_Brochure
Product Literature
Frequently Bought Together