KIMAX® Burets, Class A, Straight Bore PTFE Stopcock, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
About this item
Precision bore burets are permanently marked with an individual serial number and include a 2mm stopcock plug
Scale is a durable white ceramic enamel. Burets are designed from ASTM Specification E287, Class A requirements. Delivery stem of the titration buret is115 mm long and meets ASTM D664 potentiometric titration buret requirements.
Ordering information: Burets are supplied with a Certificate of Graduation Accuracy and include a KIM-KAP® dust cap.