Burets, Automatic Self-Zeroing, Dual Rotaflo® Stopcocks, ProScience
Supplier: Proscience
Certificates
About this item
Burets feature two Rotaflo stopcocks that help eliminate titration errors and never require lubrication
The Rotaflo® design ensures accuracy and contamination-free performance. Buret 87003-564 accepts 12mm O.D. tubing for side and fill arms. Buret 87003-566 accepts 14mm O.D. tubing. Both burets feature a durable white enamel scale.
Frequently Bought Together