KIMAX® Powder Funnels, Kimble Chase, DWK Life Sciencese
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Funnels feature short, wide stems and fire-polished tops and stems.
These powder funnels are made of glass.
Certifications: Designed from ASTM specification E1095, Type 4 requirements.
Specifications
- Catalog No:
- 89001-416
- 89001-422
- 89001-424
- 89001-418
- 89001-420
- Stem length:
- 35 mm
- 35 mm
- 35 mm
- 35 mm
- 35 mm
Specifications
Related Information
Certificates
Search by Lot NumberDWK Life Sciences Return to Research Promo 2021
Promotion Material
Kimble Petrochemical Glassware Brochure
Product Literature
Customers Who Bought This Also Bought