Orders LinkContactUsLinkComponent
Bravo Disposable Pipette Tips
Bravo Disposable Pipette Tips
Bravo Disposable Pipette Tips
Certificates
Bravo Disposable Pipette Tips
Bravo Disposable Pipette Tips
26 Options Available Below
Product Details & Documents
Certified on Bravo and AssayMAP for accuracy and precision.
  • Individual lot certified to meet thorough QC standards
  • Certified free of detectable DNase and RNase activity
  • Certified free of endotoxin (pyrogen) contamination
  • Certified free of adenosine triphosphate (ATP)
  • Certified free of human DNA;Certified free of protease activity
  • Certified free of heavy metals
  • Certified as sterile with a sterility assurance level (SAL) of 10-6
  • Innovative processing and packaging reduce static buildup and improve deck space utilization
Agilent offers disposable pipette tips in both 96- and 384-well formats specially designed, tested, and optimized for the Bravo Automated Liquid Handling Platform and the AssayMAP Bravo heads. All Agilent pipette tips undergo complete QC and functional performance testing for each lot.
The innovative processing and packaging of Bravo robotic tips helps address common concerns about lab disposable pipette tips used in automation, such as static buildup and deck space utilization. Additionally, the Agilent Bravo disposable pipette tips are the only pipette tips that are certified for use on Agilent Bravo Liquid Handling instrumentation, with the assurance that every tip meets our exacting accuracy and precision requirements.
Packaging: Packed as case of 50, except: 21740-202, 21740-212, 21750-202 and 21750-212 which are a case of 90.
Product Family Options
10 μl
Product Information
  • Tip style
    Volume
    For use with
    Packed
    Availability
    Price
  • 384 in rack, non sterile
    10 µl
    Compatible with Bravo 96ST or 384ST head
    50/Case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $6,530.00
    $6,530.00
    Pack of 50
       $6,530.00
    Minimum order quantity is 1
    Supplier Number: 10734-202
  • 384 in rack, sterile
    10 µl
    Compatible with Bravo 96ST or 384ST head
    50/case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $6,450.00
    $6,450.00
    Pack of 50
       $6,450.00
    Minimum order quantity is 1
    Supplier Number: 10734-212
  • 384 in rack, conductive
    10 µl
    Compatible with Bravo 96ST or 384ST head
    50/Case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $6,100.00
    $6,100.00
    Pack of 50
       $6,100.00
    Minimum order quantity is 1
    Supplier Number: 10734-302
  • 384 in rack, sterile conductive
    10 µl
    Compatible with Bravo 96ST or 384ST head
    50/Case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $6,290.00
    $6,290.00
    Pack of 50
       $6,290.00
    Minimum order quantity is 1
    Supplier Number: 10734-312
  • 384 in rack, nested conductive
    10 µl
    Compatible with Bravo 96ST or 384ST head
    90/case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $10,410.00
    $10,410.00
    Pack of 50
       $10,410.00
    Minimum order quantity is 1
    Supplier Number: 21740-202
  • 384 in rack, nested sterile conductive
    10 µl
    Compatible with Bravo 96ST or 384ST head
    90/case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $11,190.00
    $11,190.00
    Pack of 50
       $11,190.00
    Minimum order quantity is 1
    Supplier Number: 21740-212
30 μl
Product Information
  • Tip style
    Volume
    For use with
    Packed
    Availability
    Price
  • 384 in rack, non sterile
    30 µl
    Compatible with Bravo 96ST or 384ST head
    50/Case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $5,970.00
    $5,970.00
    Pack of 50
       $5,970.00
    Minimum order quantity is 1
    Supplier Number: 11484-202
  • 384 in rack, sterile
    30 µl
    Compatible with Bravo 96ST or 384ST head
    50/case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $6,130.00
    $6,130.00
    Pack of 50
       $6,130.00
    Minimum order quantity is 1
    Supplier Number: 11484-212
  • 384 in rack, conductive
    30 µl
    Compatible with Bravo 96ST or 384ST head
    50/Case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $4,552.66
    $4,552.66
    Pack of 50
       $4,552.66
    Minimum order quantity is 1
    Supplier Number: 11484-302
  • 384 in rack, sterile conductive
    30 µl
    Compatible with Bravo 96ST or 384ST head
    50/Case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $6,100.00
    $6,100.00
    Pack of 50
       $6,100.00
    Minimum order quantity is 1
    Supplier Number: 11484-312
  • 384 in rack, sterile filtered, conductive
    30 µl
    Compatible with Bravo 96ST or 384ST head
    50/Case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $7,380.00
    $7,380.00
    Pack of 50
       $7,380.00
    Minimum order quantity is 1
    Supplier Number: 11484-322
  • 384 in rack, nested conductive
    30 µl
    Compatible with Bravo 96ST or 384ST head
    90/case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $10,410.00
    $10,410.00
    Pack of 50
       $10,410.00
    Minimum order quantity is 1
    Supplier Number: 21750-202
  • 384 in rack, nested sterile conductive
    30 µl
    Compatible with Bravo 96ST or 384ST head
    90/case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $11,190.00
    $11,190.00
    Pack of 50
       $11,190.00
    Minimum order quantity is 1
    Supplier Number: 21750-212
70 μl
Product Information
  • Tip style
    Volume
    For use with
    Packed
    Availability
    Price
  • 384 in rack, non sterile
    70 µl
    Compatible with Bravo 96ST or 384ST head
    50/case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $6,000.00
    $6,000.00
    Pack of 50
       $6,000.00
    Minimum order quantity is 1
    Supplier Number: 19133-102
  • 384 in rack, sterile
    70 µl
    Compatible with Bravo 96ST or 384ST head
    50/Case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $6,130.00
    $6,130.00
    Pack of 50
       $6,130.00
    Minimum order quantity is 1
    Supplier Number: 19133-112
  • 384 in rack, sterile filtered, conductive
    70 µl
    Compatible with Bravo 96ST or 384ST head
    50/case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $5,963.02
    $5,963.02
    Pack of 50
       $5,963.02
    Minimum order quantity is 1
    Supplier Number: 19133-142
  • 384 in rack, conductive
    70 µl
    Compatible with Bravo 96ST or 384ST head
    50/case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $5,970.00
    $5,970.00
    Pack of 50
       $5,970.00
    Minimum order quantity is 1
    Supplier Number: 19133-202
  • 384 in rack, sterile conductive
    70 µl
    Compatible with Bravo 96ST or 384ST head
    50/Case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $6,100.00
    $6,100.00
    Pack of 50
       $6,100.00
    Minimum order quantity is 1
    Supplier Number: 19133-212
  • 384 in rack, wide bore conductive
    70 µl
    Compatible with Bravo 96ST or 384ST head
    50/case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $6,110.00
    $6,110.00
    Pack of 50
       $6,110.00
    Minimum order quantity is 1
    Supplier Number: 19134-002
  • 384 in rack, wide bore sterile conductive
    70 µl
    Compatible with Bravo 96ST or 384ST head
    50/Case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $6,270.00
    $6,270.00
    Pack of 50
       $6,270.00
    Minimum order quantity is 1
    Supplier Number: 19134-012
  • 384 in rack, wide bore sterile filtered, conductive
    70 µl
    Compatible with Bravo 96ST or 384ST head
    50/Case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $7,510.00
    $7,510.00
    Pack of 50
       $7,510.00
    Minimum order quantity is 1
    Supplier Number: 19134-022
250 μl
Product Information
  • Tip style
    Volume
    For use with
    Packed
    Availability
    Price
  • 96 in rack, non sterile
    250 µl
    Compatible with Bravo 96LT head
    50/case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $529.40
    $529.40
    Pack of 50
       $529.40
    Minimum order quantity is 1
    Supplier Number: 19477-002
  • 96 in rack, sterile
    250 µl
    Compatible with Bravo 96LT head
    50/Case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $826.00
    $826.00
    Pack of 50
       $826.00
    Minimum order quantity is 1
    Supplier Number: 19477-012
  • 96 in rack, wide bore
    250 µl
    Compatible with Bravo 96LT head
    50/case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $725.00
    $725.00
    Pack of 50
       $725.00
    Minimum order quantity is 1
    Supplier Number: 19477-032
  • 96 in rack, wide bore sterile
    250 µl
    Compatible with Bravo 96LT head
    50/Case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $861.00
    $861.00
    Pack of 50
       $861.00
    Minimum order quantity is 1
    Supplier Number: 19477-072
  • 96 in rack, wide bore sterile filtered
    250 µl
    Compatible with Bravo 96LT head
    50/case
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $1,380.00
    $1,380.00
    Pack of 50
       $1,380.00
    Minimum order quantity is 1
    Supplier Number: 19477-082