VWR® Graduated Cylinders, Class B, To Deliver
Supplier: VWR International
Certificates
About this item
Cylinders feature a durable white ceramic enamel scale and a yellow LDPE bumper on sizes 25 mL and larger.
- Durable white enamel scale
- Increased stability through shorter height
The 10 mL size has an enlarged, funnel-shaped top for ease of filling
Meet ASTM Specification E1272, Style 1, Class B requirements. The 100 mL size may be used for some ASTM distillation tests.
The 10 and 25 mL sizes are comparatively short to provide increased stability.
Customers Who Bought This Also Bought