Orders LinkContactUsLinkComponent
NIGHTSEA® Eclipse MicroTent™ for Fluorescence Microscopy
NIGHTSEA® Eclipse MicroTent™ for Fluorescence Microscopy
NIGHTSEA® Eclipse MicroTent™
Catalog # CA77463-336
Supplier #:  SFA-TENT
NIGHTSEA® Eclipse MicroTent™ for Fluorescence Microscopy
Catalog # CA77463-336
NIGHTSEA® Eclipse MicroTent™ for Fluorescence Microscopy
NIGHTSEA® Eclipse MicroTent™ for Fluorescence Microscopy
NIGHTSEA® Eclipse MicroTent™
Price
$1,148.40
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The patented Eclipse MicroTent™ is a unique product for fluorescence microscopy that provides local darkness around conventional laboratory stereo microscopes, while still providing easy access to the sample stage and the focus and zoom controls. Use with the NIGHTSEA® SFA stereo microscope fluorescence adapter system.
  • Opening for the microscope oculars with elastic sleeve to minimize light entry
  • Large front flap provides easy access to the sample stage and can be fastened open
  • Arm slots on sides to provide access to focus and zoom controls
  • User-customizable feedthrough patches to provide additional penetrations for camera port, power cords
  • Folds flat for storage
  • Dimensions: 46×30×50 cm (18×12×20")
Fluorescence microscopes are kept in dark rooms for good reason, as fluorescence can be weak and it can be difficult to see well if there is any ambient light. Microscopes may be on lab benches in shared spaces, near windows, or in other difficult-to-darken locations such as in the field.
The Eclipse Microtent™ creates local darkness around a microscope while still providing easy access to the sample stage and the focus and zoom controls. It is designed for stereo microscopes but could potentially be used with many varieties of compound microscopes.
Specifications
  • Description:
    NIGHTSEA® Eclipse MicroTent™
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)