ContactUsLinkComponent
Pierce™ Peroxidase IHC Detection Kit, Thermo Scientific
Pierce™ Peroxidase IHC Detection Kit, Thermo Scientific
Catalog # CAPI36000
Supplier:  Thermo Scientific
CAS Number:  
undefined
Pierce™ Peroxidase IHC Detection Kit, Thermo Scientific
Catalog # CAPI36000
Supplier:  Thermo Scientific
Supplier Number:  36000
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    ImmunoHisto™ Peroxidase Detection Kit, with HRP Substrate
  • Cat. no.:
    CAPI36000
  • Supplier No.:
    36000
Specifications
About this item
The Pierce™ Peroxidase IHC Detection Kit bundles the Pierce™ Metal Enhanced DAB Substrate Kit with all necessary components to stain, counterstain and preserve experimental results with frozen or paraffin tissue sections.
  • Fifty times more sensitive than the traditional DAB (3,3'-diaminobenzidine) method and 30 times more sensitive than other metal-intensified versions of DAB
  • Low background, high intensity, large dynamic range
  • Easy-to-use, two component system
  • Working solution stable for six hours
  • Complete kit
  • Compatible with direct or avidin-biotin-complex (ABC) probing strategies
Delivery: DAB/metal concentrate (1X), 25 mL; Universal Blocker™ blocking buffer in TBS, 25 mL; Peroxidase Suppressor, 2 × 1 mL; stable peroxide buffer, 25 mL; BupH™ tris buffered saline, 4 × 5 mL; Surfact-Amps® 2 (1% Tween®-2 Detergent), 1 mL; Harris modified hematoxylin (without acetic acid, mercury-free), 1 mL; and mounting medium, 6 mL.