Nalgene® Laboratory Bottles, Polymethylpentene, Wide Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Completely transparent, chemical-resistant plastic bottle is ideal for tissue specimens that must be shipped wet.
- Withstands temperatures from 20 to 135 °C (68 to 275 °F)
- With polypropylene closure
- Leakproof
- Autoclavable
For replacement caps, see 16219-030 series.
Note: Before autoclaving, set closure on top of bottle without engaging threads.
Customers Who Bought This Also Bought