Pierce 1-Step™ ABTS Substrate Solution, Thermo Scientific
CAPI37615
:
- Conjugate ELISA:HRP
- Description:1-Step™ ABTS Substrate Solution
- Size:250 ml
- Colour development:Green Stain
- Sensitivity:Low
- Cat. no.:CAPI37615
- Supplier No.:37615
ABTS substrate is a water-soluble peroxidase substrate that yields a measurable green end product for use in ELISA methods.
- Green product has two major absorbance peaks, 410 nm and 65 0nm