Order Entry
Canada
ContactUsLinkComponent
Proteasome Substrate III, Fluorogenic, Calbiochem®
Proteasome Substrate III, Fluorogenic, Calbiochem®
  CA80053-860
 :  
Proteasome Substrate III, Fluorogenic, Calbiochem®
  CA80053-860
 :  539142-5MG
 :  

 

  • Description:
    Proteasome Substrate III, Fluorogenic
  • Size:
    5 mg (0.0001 oz.)
  • Form:
    White amorphous solid
  • Purity:
    98% by HPLC
  • Solubility:
    DMSO
  • Cat. no.:
    CA80053-860
  • Supplier No.:
    539142-5MG

 

 

Fluorogenic proteasome substrate that also acts as a substrate for calpain, chymotrypsin, and ingensin.

Excitation maximum: ~380nm. Emission maximum: ~460nm.