Granzyme B Substrate II, Fluorogenic, Calbiochem®, Millipore®
Supplier: MilliporeSigma
Certificates
About this item
A fluorogenic substrate for the detection of granzyme B and caspase-8 activity
Reaction can be monitored visually or quantitatively by a blue to green shift in fluorescence upon cleavage of the AFC fluorophore. Excitation maximum: ~400nm; emission maximum: ~505nm.
Specifications
- Form:White lyophilized solid
- Purity:97% by HPLC
- Solubility:DMSO
- Molecular weight:821.8
Specifications
Frequently Bought Together