Order Entry
Canada
ContactUsLinkComponent
Caspase-1 Substrate VI, Fluorogenic, Calbiochem®
Caspase-1 Substrate VI, Fluorogenic, Calbiochem®
Catalog # CA80055-026
Supplier:  MilliporeSigma
CAS Number:  
Caspase-1 Substrate VI, Fluorogenic, Calbiochem®
Catalog # CA80055-026
Supplier:  MilliporeSigma
Supplier Number:  688225-5MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Caspase-1 Substrate VI, Fluorogenic
  • Size:
    5 mg (0.0001 oz.)
  • Form:
    White to off-white solid
  • Purity:
    98% by HPLC
  • Solubility:
    DMSO
  • Cat. no.:
    CA80055-026
  • Supplier No.:
    688225-5MG

Specifications

About this item

Fluorogenic caspase-1 substrate

Reaction can be monitored visually or quantitatively by a blue to green shift in fluorescence upon cleavage of the AFC fluorophore. Excitation maximum: ~400nm; emission maximum: ~505nm.