VITLAB® Short-Form Graduated Cylinders, Polypropylene, BrandTech
Supplier: Brandtech
Certificates
About this item
Made of translucent, chemically-resistant polypropylene.
- Short form for stability
- Molded graduations
Short-form design increases stability to help prevent spills. Circular base.
Specifications
- Catalog No:
- 83008-850
- 83008-846
- 83008-842
- 83008-858
- 83008-838
- 83008-854
- Form:
- Short
- Short
- Short
- Short
- Short
- Short