VITLAB® Graduated Cylinders, Polypropylene, Class B, BrandTech
Made of polypropylene with ring-molded graduations at major increments to simplify measurements.
- Break-resistant
- High translucency
- Polypropylene for high chemical resistance
- Hexagonal base resists rolling
Class B per DIN 12681.
- Catalog No:
- 83008-874
- 83008-924
- 83008-894
- 83008-864
- 83008-934
- 83008-914
- 83008-904
- 83008-884
- Form:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
Frequently Bought Together