Order Entry
Canada
ContactUsLinkComponent
KIMAX® Single Metric Scale Graduated Cylinder, Emulsion Test, Kimble Chase
KIMAX® Single Metric Scale Graduated Cylinder, Emulsion Test, Kimble Chase
Catalog # 89001-092
undefined
KIMAX® Single Metric Scale Graduated Cylinder, Emulsion Test, Kimble Chase
Catalog # 89001-092
Supplier Number:  20011-100

Specifications

  • Graduation Interval:
    1 ml
  • Capacity:
    100 ml
  • Description:
    Single Metric Scale Graduated Cylinder
  • Form:
    Tall
  • Cat. no.:
    89001-092
  • Supplier No.:
    20011-100

Specifications

About this item

Used to determine emulsifying and demulsifying tendencies of lubricating oils (ASTM D1401)

Round base is designed to fit baths in which the cylinder is generally used. Calibrated to contain with a durable white ceramic enamel scale.