KIMAX® Single Metric Scale Graduated Cylinders, Class B, Low Form, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Calibrated "to deliver." Cylinders are made of sturdy borosilicate glass and feature two pour spouts
Bodies have a larger diameter with lower overall height than comparable standard cylinders, providing greater stability and decreased breakage. Scale is durable white ceramic enamel.
Specifications
- Catalog No:
- 89001-200
- 89001-202
- Form:
- Short
- Short