Nalgene® Packaging Bottles, HDPE, Narrow-Mouth, with Screw Caps, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These economical, break-resistant bottles are made from high-density polyethylene.
- Available in natural or amber colors
- Guaranteed leakproof
Natural sample bottles allow easy viewing of liquid level. Amber bottles reduce UV light transmission for protection of light-sensitive liquids.
Ordering information: Supplied with polypropylene screw caps.
Related Information
Certificates
Search by Lot Number16059-550_1253603 16059-552_1266529 16059-556_1252459 TXL16059548GPI_1252194
Conformity declaration
Customers Who Bought This Also Bought