Orders LinkContactUsLinkComponent
Face Shield, 701.02
Face Shield, 701.02
Face Shield, 701.02
Certificates
Face Shield, 701.02
Face Shield, 701.02
2 Options Available Below
Product Details & Documents
The 701.02 series face shield is made from clear acetate, and features an anti-fog lens treatment.
  • Anti-fog acetate shield
  • Adjustable elastic head band
  • Shield is inclinable up to 90°
  • Clear/gray frame, clear lens
Designed with an interchangeable shield, the 701.02 series face shield can be worn with prescription glasses and protective masks. This protective device is certified according to EN 166, level 3 protection (EU) against droplets and splashes of liquids.
Ordering information: Anti-fog acetate replacement shields are available separately.
Product Family Options
Product Information
  • Lens tint
    Availability
    Price
  • 77393-772
    701.02 face shield
    701.02 face shield
    Clear
    Log in for availability
    Log in for availability
    Each
    Each - $64.36
    $64.36
    Each
       $64.36
    Supplier Number: MDU7002S
  • 77393-782
    Replacement face shield
    Replacement face shield
    Clear
    Log in for availability
    Log in for availability
    Each
    Each - $40.29
    $40.29
    Each
       $40.29
    Supplier Number: MDU7012S