Specimen Transport Bag with Pocket, Nasco*
Catalog #: CA11216-783
Supplier: Nasco
Specifications
- W×L:152×230 mm (6×9")
- Description:Non sterile transport bag with biohazard symbol
- Cat. no.:CA11216-783
- Supplier No.:B01253WA
Specifications
About this item
Unbreakable 24 oz.
Polyethylene bag with puncture-proof tabs to ensure leakproof protection. Outside pouch separates paperwork from specimen. Biohazard symbol displayed on non sterile bag. Also available as a sterile bag without symbol.
Customers Who Bought This Also Bought