ContactUsLinkComponent
Eisco Polypropylene Test Tube Basket with Lid
  470355-734
 :  Wards
undefined
Eisco Polypropylene Test Tube Basket with Lid
  470355-734
 :  Wards
 :  CH0707H
  • Color:
    White
  • Description:
    Test tube basket with lid
  • Cat. no.:
    470355-734
  • L×D×H:
    230×230×230 mm (9×9×9")
  • Weight:
    0.5 lbs
The lidded test tube basket is ideal for securely holding and organizing test tubes in the lab.
  • Autoclavable
  • Unbreakable
  • Non-corrosive
The test tube basket is made of heavy duty polypropylene. The basket features a lid, which provides additional security of the contained apparatus.