Nalgene® Large Size Funnels, Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Ideal for transferring liquids
Funnels have both internal and external ribbing for air release. High-density polyethylene.
Frequently Bought Together