ContactUsLinkComponent
Safety Glasses, Adult PC
Safety Glasses, Adult PC
  470354-652
undefined
Safety Glasses, Adult PC
  470354-652
 :  UNHS2004
  • Description:
    Safety glasses, Adult PC
  • Cat. no.:
    470354-652
  • Frame Material:
    Nylon
  • Lens Material:
    PC
Protect your eyes over your glasses.
  • Strong polycarbonate lenses
  • Fits over personal spectacles
  • Adjustable nylon frames
  • Meets ANSI Z87.1 standards
These high-quality safety glasses are ideal for protection against dust and impact. The frames fit comfortably over glasses and fit many face sizes and shapes. The impact-resistant lenses are made from durable polycarbonate.