ContactUsLinkComponent
Baseline® Fingertip Pulse Oximeter, Standard
Baseline® Fingertip Pulse Oximeter, Standard
Catalog # 470355-162
undefined
Baseline® Fingertip Pulse Oximeter, Standard
Catalog # 470355-162
Supplier Number:  12-1926
Specifications
  • Description:
    Pulse oximeter, standard
  • Cat. no.:
    470355-162
Specifications
About this item
Pulse oximeter measures and displays real-time pulse and SpO₂ level.
  • Suitable and accurate for both adult and pediatric use
  • Sturdy enough for hospitals and clinics as well as for in-home patients
  • Terrific for non-invasive spot checks of the blood oxygen level
Fingertip Pulse Oximeter is a portable, compact, non-invasive measuring tool used to determine the amount of oxygen the body is carrying. Attaching painlessly to a fingertip, it sends two wavelengths of light through the finger to measure pulse rate and how much oxygen is in the system. At the same time it's measuring the blood oxygen level, the oximeter also measures the pulse rate. Results are shown on a bright LED display that can change orientation from vertical to horizontal and back with each push of the power button.
Note, this device is not to be used for continuous monitoring. Built solidly, the pulse oximeter won't require routine calibration or maintenance other than the occasional replacement of batteries. Prior to and after each use, wipe down the silicone area that fingers touch inside the unit with 70% isopropyl alcohol (rubbing alcohol).
Ordering information: Supplied complete with Oximeter, lanyard, two AAA batteries, and illustrated instruction guide.