Order Entry
Canada
Orders LinkContactUsLinkComponent
BioXp® 3250 Systems

BioXp® 3250 Systems

BioXp® 3250 system
Catalog # 77461-216
Supplier:   TELESIS BIO
Supplier #:  BX3250-01
BioXp® 3250 Systems
Catalog # 77461-216
Supplier:  TELESIS BIO
BioXp® 3250 Systems
BioXp® 3250 Systems

BioXp® 3250 system

Price
Inquire For Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

The next generation high-throughput synthetic biology workstation for accelerating discovery.

  • Gene synthesis: Custom high-fidelity gene fragments
  • mRNA: Custom
  • Synthetic mRNA
  • DNA cloning: Custom DNA fragments, up to 7,000 bp, cloned into any vector
  • DNA amplification: Cell-free DNA amplification
  • Libraries: Custom variant DNA libraries

The BioXp™ system has empowered researchers worldwide to improve productivity, reduce turnaround time, and have complete workflow control. The solution enables synthesis of DNA, mRNA and variant libraries from digital sequence in just days and requires no prior knowledge or expertise to operate.

Researchers can securely submit sequences through the myBioXperience™ portal without risking any transfer of proprietary vector information and build sequences in their own lab using push-button automation on their own schedule.

Certifications: NRTL.

Accessories information: Power input voltage: 100 to 240 V
Power input current: 8.3 A Max
Operating temperature range: 16...40 °C
Weight: 63.4 kg (139.8 US lbs)
Dimensions (W×D×H): 69×77×53 cm (27×30×21)"

Delivery information: Expected shipping: 40 days.

Specifications
  • Description:
    BioXp® 3250 system
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Service annual offline BioXp® 3250
    Service annual offline BioXp® 3250
    77461-218
    Log in for availability
    Each

Product Information

  • Catalog #
    Availability
    Price
  • Installation BioXp® 3250 system
    Installation BioXp® 3250 system
    77461-220
    Log in for availability
    Each

Product Information

  • Catalog #
    Availability
    Price
  • Training BioXp® 3250
    Training BioXp® 3250
    77461-222
    Log in for availability
    Each

Product Information

  • Catalog #
    Availability
    Price
  • Service annual online 3250 BioXp®
    Service annual online 3250 BioXp®
    77506-802
    Log in for availability
    Each

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.