Order Entry
Canada
Orders LinkContactUsLinkComponent
Tetramethylammonium hydroxide 10% in aqueous solution, BAKER ANALYZED®, J.T.Baker®

Tetramethylammonium hydroxide 10% in aqueous solution, BAKER ANALYZED®, J.T.Baker®

Glass bottle, amber, narrow mouth in single shipper, 500 ml
Catalog # CAJTV643-7
Toxic
Corrosive
Supplier #:  BAKRV643-07
Tetramethylammonium hydroxide 10% in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Catalog # CAJTV643-7
CAS Number:  75-59-2
MOC:  Non-RegulatedCertificates
Tetramethylammonium hydroxide 10% in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Tetramethylammonium hydroxide 10% in aqueous solution, BAKER ANALYZED®, J.T.Baker®

Glass bottle, amber, narrow mouth in single shipper, 500 ml

CAS Number:  75-59-2
MOC:  Non-Regulated
Price
Inquire For Price
Log in for availability
Each (500ml) Case of 12 (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:75-59-2
  • Formula:C₄H₁₃NO
  • MDL Number:MFCD00008280
  • Molecular Weight:91.15
  • Boiling Pt:110 °C (1013 hPa)
  • Density:1.02 g/cm³ (20 °C)
  • ADR:8,II
  •  :Management of Change (MOC) category = N
  •  :Density: 1.016 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:200-882-9
  • UN:1835
Specifications

Specification Test Results

  • Assay ((CH₃)₄NOH):
    ≥9.8 %
  • Residue after Ignition:
    ≤0.020 %
  • Ammonia and Other Amines:
    Passes Test

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth in single shipper
  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.