Order Entry
Canada
Orders LinkContactUsLinkComponent
PowrTrol Power Controller, Glas-Col®

PowrTrol Power Controller, Glas-Col®

Power Controller
Catalog # 33797-414
Small Business Enterprise
Supplier:   Glas-Col
Supplier #:  104APL120
PowrTrol Power Controller, Glas-Col®
Catalog # 33797-414
Supplier:  Glas-Col
Small Business Enterprise
PowrTrol Power Controller, Glas-Col®
PowrTrol Power Controller, Glas-Col®

Power Controller

Price
$704.36
Log in for availability
Each
Product Details & Documents

A solid state, proportional voltage power control that provides precise manual control of heating mantles, tapes, cords, and other resistive loads.

Output range is adjustable from 3–100% of rated voltage

Supplied with lighted on/off switch, three-wire load receptacle, and three-wire line cord with molded plug.

May be mounted on framework or used on bench top.

Specifications
  • Description:
    Power Controller
  • Cat. No.:
    33797-414
  • Dimensions:
    10.1W x 10.1D x 8.6H cm (4 x 4 x 33/8")
  • Electrical:
    120V, 1200W, 10A