Order Entry
Canada
ContactUsLinkComponent
2-Propanol ≥99.5% VLSI, J.T.Baker®
Flammable
Irritant
2-Propanol ≥99.5% VLSI, J.T.Baker®
Catalog # CAJT5373-5
CAS Number:  
MOC:  RegulatedCertificates
undefined
Flammable
Irritant
2-Propanol ≥99.5% VLSI, J.T.Baker®
Catalog # CAJT5373-5
Supplier Number:  BAKR5373-05
CAS Number:  
MOC:  RegulatedCertificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Poly bottle, pressure bottom, bagged
  • Size:
    8 pt.
  • Cat. no.:
    CAJT5373-5
  • Supplier No.:
    5373-05

Specifications

Specification Test Results

  • Assay (CH₃CHOHCH₃):
    ≥ 99.5 %
  • Color (APHA):
    ≤ 10
  • Residue after Evaporation:
    ≤ 4 ppm
  • Solubility in H₂O:
    Passes Test
  • Water (H₂O)(by Karl Fischer titrn):
    ≤ 0.05 %
  • Acidity (µeq/g):
    ≤ 0.2
  • Alkalinity (µeq/g):
    ≤ 0.1
  • Chloride (Cl):
    ≤ 0.2 ppm
  • Phosphate (PO₄):
    ≤ 0.3 ppm
  • Trace Impurities - Aluminum (Al):
    ≤ 50.0 ppb
  • Arsenic and Antimony (as As):
    ≤ 10.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤ 20.0 ppb
  • Trace Impurities - Boron (B):
    ≤ 10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤ 20.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤ 50.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤ 20.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤ 20 ppb
  • Trace Impurities - Copper (Cu):
    ≤ 10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤ 50 ppb
  • Trace Impurities - Germanium (Ge):
    ≤ 50.0 ppb
  • Trace Impurities - Gold (Au):
    ≤ 20 ppb
  • Heavy Metals (as Pb):
    ≤ 200.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤ 50.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤ 20.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤ 50.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤ 20 ppb
  • Trace Impurities - Manganese (Mn):
    ≤ 20.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤ 10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤ 100 ppb
  • Trace Impurities - Silicon (Si):
    ≤ 50 ppb
  • Trace Impurities - Silver (Ag):
    ≤ 20.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤ 100.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤ 20.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤ 100 ppb
  • Trace Impurities - Titanium (Ti):
    ≤ 20 ppb
  • Trace Impurities - Zinc (Zn):
    ≤ 50 ppb
  • Particle Count - 0.5 µm and greater:
    ≤ 100 par/ml
  • Particle Count - 1.0 µm and greater:
    ≤ 8 par/ml

Specification Test Results

About this item

 :
 :60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
 :Management of Change (MOC) category = R
REACH:01-2119457558-25
 :GHS02
 :GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674