About this item
Used in many laboratory and plant environments, these Infrared Thermometers provide non-contact °F or °C (0.1 resolution) temperature measurement of any liquid, solid, or semi-solid.
- Simple operation
- Backlit readout
- Offered in 3 different models
- Calibration points: 100, 200 °C
The -33/220°C thermometer has a 6:1 DTS, adjustable emissivity for greater measuring capabilities, max/min memory, high/low alert, error message indicators, lock mode for continuous temperature monitoring, laser sighting, and backlit readout
The two following models can be used in high temperature range applications when the user needs to be a safe distance from the target being measured. They feature max/min memory, high/low alert, laser sighting, lock function for continuous measurement, average and differential, backlit readout, and auto shut off.
The -60/500°C thermometer measures both non–contact and contact temperatures with adjustable emissivity and has an 12:1 DTS. It includes a built-in thermocouple port that can accommodate any type K thermocouple with a mini-connector when direct contact measurement is required (thermocouple sold separately).
The -20/537°C thermometer offers a 12:1 DTS, a 12-dot outline laser sighting, and it is easy to store (carrying case included).