SP Wilmad-LabGlass Volumetric Serialized Flasks with Stopper, Class A, SP Industries
Supplier: SP Industries
Certificates
About this item
Glass quality, calibration lines, inscription and appearance of these borosilicate glass flasks are maintained according to the former NBS circular 602.
- They have a marking spot in addition to a serial number
- Calibrated “to contain” (TC) at 20 °C
- Supplied with stopper
Frequently Bought Together