Sodium thiosulfate 0.20 N (N/5), StandARd® volumetric solution, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
CAS Number:7772-98-7
:
Formula:Na₂S₂O₃
MDL Number:MFCD00003499
:158.11 g/mol
Density:1.06 g/cm³
:Management of Change (MOC) category = N
Storage Temperature:Ambient
EINECS:231-867-5
Specification Test Results
- Normality:0.199 - 0.201
Specification Test Results
Related Information
Certificates
Search by Lot Number5531 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_MACR5531
SDS
Frequently Bought Together