D-(-)-Sorbitol ≥91.0%, BAKER ANALYZED®, Biochemical Reagent
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
D-(-)-Sorbitol ≥91.0%, BAKER ANALYZED®, Biochemical Reagent
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Lot analysis on label.
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
- CAS Number:50-70-4
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00004708
- Molecular Weight:182.17 g/mol
- Merck Index:13,08797
- Boiling Pt:295 °C (1013 hPa)
- Melting Pt:94…98 °C
- Density:1.524 g/cm³ (20 °C)
- :Management of Change (MOC) category = N
- Flash Pt:>100 °C
- Storage Temperature:Ambient
- EINECS:200-061-5
1 Options Available Below
Product Details & Documents
Lot analysis on label.
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
- CAS Number:50-70-4
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00004708
- Molecular Weight:182.17 g/mol
- Merck Index:13,08797
- Boiling Pt:295 °C (1013 hPa)
- Melting Pt:94…98 °C
- Density:1.524 g/cm³ (20 °C)
- :Management of Change (MOC) category = N
- Flash Pt:>100 °C
- Storage Temperature:Ambient
- EINECS:200-061-5
Documents
Specifications for Product Family
Specification Test Results
- Assay (C₆H₁₄O₆):≥91.0 %
- Ash (sulfated):≤0.1 %
- Loss on Drying at 80°C (in vacuo):≤1.0 %
- Reducing Sugars:≤0.3 %
- Total Sugars:≤1.0 %
- Chloride (Cl):≤0.005 %
- Sulfate (SO₄):≤0.01 %
- Arsenic (As):≤0.0003 %
- Heavy Metals (as Pb):≤0.001 %
Product Family Options
Product Information
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



