SP Wilmad-LabGlass Kohlrausch Volumetric Flasks, Class A, SP Industries
Supplier: SP Industries
Certificates
About this item
Calibrated ""to contain"" at 20°C, the upper neck of these borosilicate glass flasks are enlarged to permit easy introduction of solids
Each flask is provided with a marking spot in addition to a serial number.
Ordering information: Supplied with Certificate of Compliance.
Frequently Bought Together