Sodium hydroxide concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # CAJT4687-1
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:LDPE ampoule, DILUT-IT®
- Size:1 pack
- Environmentally Preferable:
- Cat. no.:CAJT4687-1
- Supplier No.:4687-01
Specifications
Specification Test Results
- Normality (by titrimetry):Passes Test
Specification Test Results
About this item
Synonyms: Caustic soda
:
:40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Density:1.327 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,II
:Management of Change (MOC) category = N
EINECS:215-185-5
UN:1824
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548