Experience optimal performance with Thermo Scientific Nunc™ EASYFlasks™ with the Nunclon™ supra surface treatment for culturing a wide variety of fastidious adherent cells.
- Consistent lot-to-lot performance
- Sterile with SAL 10⁻⁶
- Non-cytotoxic
- Non-pyrogenic
- Biocompatible per USP Class VI for raw materials
- Shelf life five years
The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers. Intended for general cell culture applications, the Nunclon™ supra surface is more hydrophilic than the Nunclon™ delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
Nunclon™ supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Thus, Nunclon™ supra cultureware is a great xeno-free solution for cell therapy research.