Order Entry
Canada
ContactUsLinkComponent
 
KIMAX® Volumetric Flasks with [ST] Polyethylene Stopper, Class B, Kimble Chase
  89002-006
undefined
KIMAX® Volumetric Flasks with [ST] Polyethylene Stopper, Class B, Kimble Chase
  89002-006
 :  28015P-100

 

  • Capacity:
    100 ml
  • Cat. no.:
    89002-006
  • ST Stopper No.:
    13
  • Tolerance:
    ±0.16 ml
  • Supplier No.:
    28015P-100

 

 

Flasks have a graduation ring blasted on the neck

Designed from ASTM specification E288, Class B requirements.

Calibrated to contain. Provided with yellow and blue closed-bottom linear, high-density polyethylene stopper to fit the [ST] neck. Enlarged stopper top protects the flask neck if tipped over.