Order Entry
Canada
ContactUsLinkComponent
Sodium hydroxide 2.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
 
Sodium hydroxide 2.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
  CAJT0390-7
 :  
     
undefined
 
Sodium hydroxide 2.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
  CAJT0390-7
 :  BAKR0390-07
 :  
     

 

  • Pack type:
    Jerrican (poly pail)
  • Size:
    19 L
  • Cat. no.:
    CAJT0390-7
  • Supplier No.:
    0390-07

 

 

  • Appearance (Clear, colorless liquid):
    Passes Test
  • Normality:
    1.9 - 2.1
  • Chloride (Cl):
    ≤5 ppm
  • ACS - Heavy Metals (as Pb):
    ≤1 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.5 ppm
  • Endotoxin Concentration (EU/mL:
    ≤2

 

 

 :
 :40 g/mol
 :109 °C (1013 hPa)
 :≤1.11 g/cm³ (20 °C)
 :Ambient
 :8,II
 :Management of Change (MOC) category = R
 :215-185-5
 :1824
 :GHS05
 :1310-73-2
 :NaOH
 :MFCD00003548