Disodium hydrogen phosphate heptahydrate 98.0-100.5%, EMPROVE® ESSENTIAL DAC, USP, SAFC®
Catalog # CA1.06574.1000
Supplier: EMD MILLIPORE # PCS CA EXCEP
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Size:1 kg
- Cat. no.:CA1.06574.1000
- Supplier No.:1.06574.1000
Specifications
Specification Test Results
- Assay (alkalimetric, calculated on dried substance):98.0 - 100.5 %
- Identity:passes test
- Appearance of solution:passes test
- Insoluble substances:≤ 0.4 %
- pH-value (5 %; water):8.7 - 9.3
- Sodium Dihydrogene phospate (NaH₂PO₄):passes test
- Chloride (Cl):≤ 0.0200 %
- Sulfate (SO₄):≤ 0.0500 %
- Heavy metals (as Pb):≤ 0.0010 %
- As (Arsenic):≤ 0.0002 %
- Fe (Iron):≤ 0.0020 %
- Residual solvents (ICH Q3C):excluded by manufacturing process
- Reducing substances:passes test
- Loss on drying (130 °C):45.0 - 49.0 %
Specification Test Results
About this item
:
:268.07 g/mol
Melting Pt:48 °C
Density:1.7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-448-7
CAS Number:7782-85-6
Formula:HNa₂O₄P·7H₂O
MDL Number:MFCD00149180