Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ Red)
Supplier: AAT BIOQUEST INC
Certificates
About this item
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
Caution: For research use only.
Type: Secondary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: mFluor™ Red
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77021-096
- 77023-494
- Antigen symbol:
- CD3e
- CD3e
- Antigen name:
- CD3e antigen
- CD3e antigen
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- mFluor™ Red
- mFluor™ Red
- Clone:
- SK7
- SK7
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Purification:
- Affinity purification
- Affinity purification
- Storage temperature:
- Refrigerated (2...8 °C); Minimize light exposure
- Refrigerated (2...8 °C); Minimize light exposure
- Shipping temperature:
- 20
- 20
Specifications
Frequently Bought Together