Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 790)
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
: For research use only.
Type: Secondary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: iFluor® 790
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Catalog No:
- 77024-184
- 77024-964
- Antigen symbol:
- CD3e
- CD3e
- Antigen name:
- CD3e antigen
- CD3e antigen
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 790
- iFluor® 790
- Clone:
- SK7
- SK7
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Purification:
- Affinity purification
- Affinity purification
- Storage temperature:
- Refrigerated (2...8 °C); Minimize light exposure
- Refrigerated (2...8 °C); Minimize light exposure
- Shipping temperature:
- 20
- 20
Frequently Bought Together