Order Entry
Canada
ContactUsLinkComponent
 
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 488)
 
undefined
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 488)

 

  • Catalog No:
  • 77022-988
  • 77021-946
  • Antigen symbol:
  • CD3e
  • CD3e
  • Antigen name:
  • CD3e antigen
  • CD3e antigen
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • iFluor® 488
  • iFluor® 488
  • Clone:
  • UCHT1
  • UCHT1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Purification:
  • Affinity purification
  • Affinity purification
  • Storage temperature:
  • Refrigerated (2...8 °C); Minimize light exposure
  • Refrigerated (2...8 °C); Minimize light exposure
  • Shipping temperature:
  • 20
  • 20

 

Frequently Bought Together